Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide StandAR, Macron Fine Chemicals™

Catalog No. MKH320100 Shop All Avantor Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MK-H320-100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-H320-100 Supplier Avantor Supplier No. H32001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 1310-58-3
Assay Percent Range 10%
Molecular Formula HKO
MDL Number MFCD00003553
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
Quantity 100 mL
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES [OH-].[K+]
IUPAC Name potassium hydroxide
Molecular Weight (g/mol) 56.11
PubChem CID 14797
ChEBI CHEBI:32035
Packaging Poly
Chemical Name or Material Potassium Hydroxide, (w/v) Aqueous
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.