Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, Technical, ≥85%, Solstice

Catalog No. 60044579 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
5 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-579 5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-044-579 Supplier Solstice Supplier No. 061035KG
Add to Cart
Add to Cart

Technical,≥85%, powder

  • Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%
  • Variation in color and physical form is possible
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

pH 14 at 20°C
Color White
Boiling Point 1327°C
Melting Point 360°C
Density 2.04 g/cu.cm
Quantity 5 kg
Percent Purity ≥85%
Grade Technical
Packaging Poly Bottle
Vapor Pressure 1mmHg (719°C)
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.