Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium indigotrisulfonate ozone-scavenging reagent | 67627-18-3 | MFCD00013160 | 5G
Catalog No. NC1226938
Encompass
Potassium indigotrisulfonate ozone-scavenging reagent | Purity: >60% (HPLC) | Mol Wt: 616.72 | 67627-18-3 | MFCD00013160 | 5G
Catalog No. NC1226938 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2340875G
Potassium indigotrisulfonate ozone-scavenging reagent | Purity: >60% (HPLC) | Mol Wt: 616.72 | 67627-18-3 | MFCD00013160 | 5G