Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Indigotrisulfonate, Sigma-Aldrich

Catalog No. 01258986 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-986 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-986 Supplier MilliporeSigma Supelco Supplier No. 2340875G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ozone-scavenging reagent used for the colorimetric determination of ozone concentrations in water and atmospheric applications.

Ozone-scavenging reagent that reacts rapidly with ozone.

Potassium indigotrisulfonate has been used:

  • in colorimetric method of measurement of atmospheric ozone
  • for measuring residual aqueous ozone by gas diffusion flow injection analysis.
  • to determine O3 concentration in water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67627-18-3
Molecular Weight (g/mol) 616.72 g/mol

Specifications

Concentration >60%
Color Dark Blue
Quantity 5 g
Physical Form Crystals or Chunks
Percent Purity >60%
Grade Not Available
Chemical Name or Material Potassium indigotrisulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.