Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodate, 0.00564 N, 200 ppm as Cl2, Ricca Chemical
SDP

Catalog No. 627016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
627016 500 mL Amber Glass Bottle
6270-32 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 627016 Supplier Ricca Chemical Company Supplier No. 627016
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

APHA for Chlorine Testing

Chemical Identifiers

Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
PubChem CID 23665710

Specifications

Color Colorless
CAS 7758-05-6
CAS Min % 98.0, 0.02
CAS Max % 100.0, 0.02
SMILES [O-]I(=O)=O.[K+]
IUPAC Name potassium;iodate
Equivalence 1 mL = 0.2 mg Cl
Grade Laboratory
Packaging Amber Glass Bottle
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.00564Normal
Chemical Name or Material Potassium Iodate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.