Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodate, 0.00564 N, 200 ppm as Cl2, Ricca Chemical
SDP

Catalog No. 627032 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6270-32 1 L Amber Glass Bottle
627016 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6270-32 Supplier Ricca Chemical Company Supplier No. 627032
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

APHA for Chlorine Testing

Chemical Identifiers

Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M

Specifications

CAS 5-6-7758
SMILES [K+].[O-][I](=O)=O
IUPAC Name potassium iodate
Grade Laboratory
Packaging Amber Glass Bottle
Quantity 1 L
Chemical Name or Material Potassium Iodate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.