Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodate, 0.01587 N, 1 mL = 1 mg Na2SO3, Ricca Chemical
SDP

Catalog No. 627232 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6272-32 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6272-32 Supplier Ricca Chemical Company Supplier No. 627232
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ASTM for Sulfite Testing

Specifications

Volume (Metric) 1 L
Packaging Amber Glass Bottle
Quantity 1 L
Type Potassium Iodate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.