Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium iodate, 0.1N Standardized Solution

Catalog No. AA35586K2
Click to view available options
Quantity:
1 L
4 L

Catalog No. AA35586K2

Supplier: Thermo Scientific Chemicals 035586K2

Only null left
Add to Cart

CAS: 7758-05-6 | IKO3 | 214.00 g/mol

Potassium iodate 0.1N solution is used as a standard solution in volumetric titrations. It is extensively used in analytical chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
MDL Number MFCD00011406
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
PubChem CID 23665710
IUPAC Name potassium;iodate
SMILES [O-]I(=O)=O.[K+]

Specifications

Color Colorless
CAS 7758-05-6, 7732-18-5
Concentration 0.1N
Solubility Information Miscible with water,potassium iodide solution and sulfuric acid.
Formula Weight 214
Odor Odorless
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Potassium iodate, Standardized Solution
EINECSNumber 231-831-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.