Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium iodate ACS reage100G
Catalog No. 502897238
Encompass
KIO3 can be used as a substitute of KI in radiation protection. A kinetic study of thermal degradation of KIO3 by³-rays suggests that rate of decomposition increases while activation energy decreases upon irradiation.
Catalog No. 50-289-7238 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 215929100G
KIO3 can be used as a substitute of KI in radiation protection. A kinetic study of thermal degradation of KIO3 by³-rays suggests that rate of decomposition increases while activation energy decreases upon irradiation.