Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodate-Iodide, 0.0125 Normal Standardized, Reagents

Catalog No. 01336422
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-422 4 L
01-336-424 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-422 Supplier Reagents Holdings Llc Supplier No. C72860104C
Only null left
Add to Cart
Add to Cart

Potassium Iodate-Iodide, 0.0125 Normal Standardized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Specifications

Chemical Name or Material Potassium Iodate-Iodide, 0.0125 Normal Standardized
Boiling Point 100°C
CAS 7732-18-5, 7681-11-0, 7681-11-0
Color Colorless
Physical Form Liquid
Quantity 4 L
Molecular Formula KIO3/KI/H2O
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.001
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.