Learn More
Potassium Iodate-Iodide, 0.0125 Normal Standardized, Reagents
Specifications
Specifications
| Chemical Name or Material | Potassium Iodate-Iodide, 0.0125 Normal Standardized |
| Boiling Point | 100°C |
| Color | Colorless |
| CAS | 7732-18-5, 7681-11-0, 7681-11-0 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Molecular Formula | KIO3/KI/H2O |
| Grade | Reagent |
| Packaging | Glass Bottle |
| Specific Gravity | 1.001 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.