Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodate-Iodide, 0.100 N (N/10), Ricca Chemical
SDP

Catalog No. 628516 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
628516 500 mL Amber Glass Bottle
6285-32 1 L Amber Glass Bottle
6285-1 4 L Amber Glass Bottle
6285-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 628516 Supplier Ricca Chemical Company Supplier No. 628516
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Potassium Iodate-Iodide
Color Colorless
CAS 7732-18-5, 7681-11-0, 7758-05-6, 1310-73-2
CAS Min % 95.0, 2.63, 0.36, 0.1
CAS Max % 99.0, 2.74, 0.37, 0.1
Synonym dihydrogen oxide, dihydrogen monoxide
Grade Laboratory
Packaging Amber Glass Bottle
pH Slightly Alkaline
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.100Normal
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.