Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, 0.1 Normal (N/10), Unstandardized, Reagents

Catalog No. 01334261
Change view
Click to view available options
Quantity: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-334-261 Quantity: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-261 Supplier Reagents Holdings Llc Supplier No. CP1895001C
Add to Cart
Add to Cart

Potassium Iodide, 0.1 Normal (N/10), Unstandardized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Potassium Iodide, 0.1 Normal (N/10), Unstandardized
Boiling Point 100°C
CAS 7732-18-5, 7681-11-0
Color Colorless to Light Yellow
Physical Form Liquid
Quantity 1 L
Molecular Formula KI/H2O
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.0113
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.