Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, 0.5% (w/v), Ricca Chemical
SDP

Catalog No. 628716 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6287-16 500 mL Amber Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6287-16 Supplier Ricca Chemical Company Supplier No. 628716
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7681-11-0 , 7732-18-5
Molecular Formula IK
Molecular Weight (g/mol) 166.00
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium iodide
SMILES [K+].[I-]

Specifications

Percent Purity 0.5%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.5% (w/v) Aqueous Solution
Packaging Amber Poly Bottle
Treatment(s) Stabilized
Quantity 500 mL
Chemical Name or Material Potassium Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.