Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, 1.5% (w/v), Ricca Chemical
SDP

Catalog No. R628750020F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
20 L
Packaging:
Cubitainer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R628750020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R628750020F Supplier Ricca Chemical Company Supplier No. R628750020F
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Color Colorless
Physical Form Liquid
Quantity 20 L
Assay Percent Range 1.5% (w/v)
Percent Purity 1.5% W/V
Grade Laboratory
Odor Odorless
Packaging Cubitainer
Chemical Name or Material Potassium Iodide, Aqueous Solution
Recommended Storage 15°C - 30°C (59°F - 86°F)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.