Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, 1 Normal (16.6% w/v), Reagents

Catalog No. 01336429
Change view
Click to view available options
Quantity:
10 L
Packaging:
Cubitainer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-336-429 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-429 Supplier Reagents Holdings Llc Supplier No. C728600010F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potassium Iodide, 1 Normal (16.6% w/v), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7681-11-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Boiling Point 101°C
Color Colorless to Light Yellow
Physical Form Liquid
Quantity 10 L
Percent Purity 16.6%
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.12
Chemical Name or Material Potassium Iodide, 1 Normal (16.6% w/v)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.