Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Potassium Iodide 10% w/v Solution (4L)
SDP

Catalog No. NC9520726
Encompass_Preferred

Potassium Iodide 10% w/v Solution (4L)

Catalog No. NC9520726 Supplier Aqua Solutions Supplier No. 74004LC
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potassium Iodide 10% w/v Solution (4L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.