Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Potassium Iodide
SCI_ED

Catalog No. S25493D Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Lab Grade
Reagent Grade
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25493D 500 g Lab Grade
S25493 25 g Reagent Grade
S25493C 100 g Lab Grade
S25493A 100 g Reagent Grade
S25493B 500 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S25493D
Add to Cart
Edge
Add to Cart

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Color White
Quantity 500 g
Assay Percent Range >95%
Linear Formula KI
Formula Weight 166.01
Packaging Lab Grade
Density 2.165g/mL
Chemical Name or Material Potassium Iodide
Safety handling Check Alert storage code Red
ShelfLife 1 year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.