Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, 2% Buffered EPA Reference Method, Reagents

Catalog No. 01334242
Change view
Click to view available options
Quantity: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-334-242 Quantity: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-242 Supplier Reagents Holdings Llc Supplier No. CP1970001C
 Restricted Item
Add to Cart
Add to Cart

Potassium Iodide, 2% Buffered EPA Reference Method, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Potassium Iodide, 2% Buffered EPA Reference Method
Boiling Point 100°C
CAS 7732-18-5, 7681-11-0, 7778-77-0
Color Colorless to Pale Yellow
pH 7
Physical Form Liquid
Quantity 1 L
Molecular Formula KI/H2O/NaHCO3
Percent Purity 2%
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.0189
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.