Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, 20% (w/v) Aqueous Solution, Spectrum™ Chemical
SDP

Catalog No. 18612772 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-772 20 L Amber Glass Bottle
18-612-769 500 mL Amber Glass Bottle
18-612-770 1 L Amber Glass Bottle
18-612-771 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-612-772 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P43320LT
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7681-11-0 , 7732-18-5

Specifications

Physical Form Solution
Quantity 20 L
Percent Purity 20% ±0.4%
Grade Ungraded
Packaging Amber Glass Bottle
Chemical Name or Material Potassium Iodide, 20 Percent (w/v) Aqueous Solution

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.