Learn More
Electron Microscopy Sciences Potassium Iodide 50 GR
SDP
Catalog No. 5028044
Encompass_Preferred
(dodeca-Molybdophosphoric Acid)
CAS #7681-11-0 KI F.W. 166.00
Used for staining synapses. Pfenninger, K., Sandri, C., Albert, K. and Eugster, C.H. (1969). Contribution of the problem of structural organization of the presynaptic area. Brain Res., 12:10.
Catalog No. 50-280-44 Supplier Electron Microscopy Sciences Supplier No. 20158
Add to Cart
Add to Cart
(dodeca-Molybdophosphoric Acid)
CAS #7681-11-0 KI F.W. 166.00
Used for staining synapses. Pfenninger, K., Sandri, C., Albert, K. and Eugster, C.H. (1969). Contribution of the problem of structural organization of the presynaptic area. Brain Res., 12:10.