Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, Briquettes p.a., Thermo Scientific™

Catalog No. AC206475000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC206475000 500 g
AC206471000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC206475000 Supplier Thermo Scientific Chemicals Supplier No. 206475000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Color White
pH 6 to 8 (5% soln. at 20°C)
Physical Form Crystals and Briquettes
Quantity 500 g
Linear Formula KI
Fieser 06,227; 11,440
Merck Index 15, 7764
Formula Weight 166
Percent Purity ≥99.5% (on dried substance)
Loss on Drying 1% max.
Appearance Clear, colorless
Density 3.13
Chemical Name or Material Potassium iodide, p.a.
Show More Show Less
Hazard Category LIGHT SENSITIVE
Hazard Statement MOISTURE SENSITIVE
EINECSNumber 231-659-4
RTECSNumber TT2975000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.