Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, Certified, 2.0% (w/v) ±0.1%, LabChem™

Catalog No. LC197201 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Poly
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC197201 500 mL Amber Poly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC197201 Supplier LabChem, Inc. Supplier No. LC197201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7681-11-0 , 7732-18-5
Molecular Formula IK
Molecular Weight (g/mol) 166.003
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Color Colorless
CAS Max % 98, 2
Physical Form Liquid
Quantity 500 mL
Linear Formula KI
Solubility Information Soluble in water
Formula Weight 166
Percent Purity 2.0% ±0.1% (w/v)
Grade Certified
Identification Passes Test
Packaging Amber Poly
Specific Gravity 1.01g/mL
Decomposition Information Iodine vapor
Density 1.01g/mL
Chemical Name or Material Potassium Iodide
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature; Protect from light
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.