Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, Crystal, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18612783 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Poly Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-783 45 kg Amber Glass Bottle
18-612-777 25 g Amber Glass Bottle
18-612-778 125 g Amber Glass Bottle
18-612-779 500 g Amber Glass Bottle
18-612-780 2.5 kg Amber Glass Bottle
18-612-781 2.5 kg Poly Bottle
18-612-782 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-612-783 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P133545KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.00
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
IUPAC Name potassium iodide
SMILES [K+].[I-]

Specifications

CAS Max % 1
pH 6.0 to 9.2
Physical Form Crystalline Powder or Needles
Quantity 45 kg
Percent Purity 99%
Grade ACS/Reagent
Loss on Drying 0.002
Packaging Amber Glass Bottle
Chemical Name or Material Potassium Iodide, Crystal

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.