Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, For Arsenic, Certified, 15.0% (w/v) ±0.5% (w/v), LabChem™

Catalog No. LC197806
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC197806 10 L
LC197801 500 mL
LC197802 1 L
LC197804 4 L
LC197805 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. LC197806 Supplier LabChem, Inc. Supplier No. LC197806
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7681-11-0 , 7732-18-5
Molecular Formula IK
Molecular Weight (g/mol) 166.003
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Color Yellow
CAS Max % 85, 15
Linear Formula KI
Density 1.11g/mL
Solubility Information Soluble in water
Formula Weight 166
Percent Purity 15.0% ±0.5% (w/v)
Grade Certified
Identification Passes Test
Packaging Cubitainer™
Specific Gravity 1.11g/mL
Traceability to NIST Traceable to NIST
Viscosity 0.84cSt
Decomposition Information Iodine vapor
Quantity 10 L
Physical Form Liquid
Chemical Name or Material Potassium Iodide, For Arsenic
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes eye irritation.
Precautionary Statement GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature; Protect from light
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.