Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, Fusion Flux Additive (Crystalline or Liq.; for Sample Prepn.)
SDP

Catalog No. FF0700 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FF-0700 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FF-0700 Supplier SPEX CertiPrep Supplier No. FF0700

missing translation for 'validMainframeMsg'

Specifications

Packaging Poly Bottle
Physical Form Crystalline or Liquid
Quantity 100 g
Chemical Name or Material Potassium Iodide, Fusion Flux Additive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.