Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, Granular, FCC, 99-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18605622 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-622 45 kg Amber Glass Bottle
18-605-616 125 g Amber Glass Bottle
18-605-617 500 g Amber Glass Bottle
18-605-619 2.5 kg Amber Glass Bottle
18-605-618 6 x 500 g Amber Glass Bottle
18-605-620 4 x 2.5 kg Amber Glass Bottle
18-605-621 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-605-622 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P134445KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

P1344, 7681-11-0, KI

Spectrum™ Chemical Potassium Iodide, Granular, FCC is used as a source for dietary iodine. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.00
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
IUPAC Name potassium iodide
SMILES [K+].[I-]

Specifications

CAS Max % 1
Physical Form Granular/Lump
Quantity 45 kg
Percent Purity 99 to 101.5%
Grade FCC
Loss on Drying 0.01
Packaging Amber Glass Bottle
Chemical Name or Material Potassium Iodide, Granular
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.