Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, Granular, Reagent, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18605626 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-626 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-605-626 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1340500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P1340, 7681-11-0, KI

Spectrum™ Chemical Potassium Iodide, Granular, Reagent, ACS is refered to by the chemical formula KI and is an inorganic compound. Used as a component in film photography before the discovery of silver iodide, it is also used in the electrolyte of dye sensitized solar cells. KI is also used as an emergency blockade of radio-iodine, a common form of radiation poisoning. As an ACS grade Reagent, Spectrum Chemical manufactured potassium iodide is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.00
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
IUPAC Name potassium iodide
SMILES [K+].[I-]

Specifications

pH 6.0 to 9.2
CAS Max % 1
Quantity 500 g
Percent Purity 99%
Grade ACS
Loss on Drying 0.002
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.