Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide - Iodine Reagent, TAPPI for Starch, Ricca Chemical
SDP

Catalog No. R63167001C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R63167001C 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R63167001C Supplier Ricca Chemical Company Supplier No. R63167001C
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Potassium Iodide - Iodine Reagent
Color Brown
CAS 7732-18-5, 7681-11-0, 7553-56-2
CAS Min % 97.0, 0.73, 0.48
CAS Max % 100.0, 0.76, 0.5
Synonym dihydrogen oxide, dihydrogen monoxide
Grade Laboratory
Packaging Amber Glass Bottle
Quantity 1 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.