Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, Purified, Reagents

Catalog No. 01337098
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-098 3 kg
01-337-100 100 g
01-337-097 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-098 Supplier Reagents Holdings Llc Supplier No. C22860003A5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potassium Iodide, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.00
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
IUPAC Name potassium iodide
SMILES [K+].[I-]

Specifications

Boiling Point 1330°C
Melting Point 0°C
Color Colorless/White to Pale Yellow
Physical Form Solid
Quantity 3 kg
Percent Purity 99.7%
Grade Pure
Packaging Poly Bottle
Specific Gravity 3.12
Chemical Name or Material Potassium Iodide, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.