Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, Reagent Grade, Innovating Science™
SCI_ED

Catalog No. S000042 Shop All Aldon Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000042 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000042 Supplier Aldon Corporation Supplier No. IS25115
Add to Cart
Add to Cart

Chemical Identifiers

Molecular Weight (g/mol) 40 g/mol

Specifications

Physical Form Beads
Quantity 100 g
CAS 1310-73-2
Percent Purity 96 to 100%
Grade Laboratory
Packaging HDPE
Chemical Name or Material Sodium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.