Learn More
Potassium Iodide, Saturated at 20°C, Reagents
Specifications
Specifications
| Boiling Point | 108°C |
| Color | Colorless to Yellow |
| Physical Form | Liquid |
| Quantity | 4 L |
| Grade | Reagent |
| Packaging | Safety Coated Glass |
| Specific Gravity | 1.69 |
| Chemical Name or Material | Potassium Iodide, Saturated at 20°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.