Learn More
Bioworld Potassium Iodide Solution 10% (w/v), 500 mL
Catalog No. 50255579
Encompass_Preferred
A simple iodide salt utilized as a fluorescence quenching agent Appearance: LiquidBoiling Point : 1330 °CDensity : 3.12 g/cm3 at 20 °CMelting Point: 681 °C (lit.)RTECS Number: TT2975000
Catalog No. 50-255-579 Supplier Bioworld Supplier No. 402201311
A simple iodide salt utilized as a fluorescence quenching agent Appearance: LiquidBoiling Point : 1330 °CDensity : 3.12 g/cm3 at 20 °CMelting Point: 681 °C (lit.)RTECS Number: TT2975000