Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, TraceSELECT™, ≥99.999% (metals basis), Solstice

Catalog No. 60045831 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-831 100 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-045-831 Supplier Solstice Supplier No. 30533100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Boiling Point 1323°C (1013hPa)
Melting Point 681°C
Color Colorless
pH 6.0 to 9.0 (at 20°C)
Physical Form Crystalline
Quantity 100 g
Linear Formula KI
UN Number NONH for all modes of transport
Formula Weight 166g/mol
Percent Purity ≥99.999% (metals basis)
Grade TraceSELECT
Odor Odorless
Packaging Poly bottle
Vapor Pressure 1mmHg (745°C)
Density ∽3.12g/cu.cm at 20°C
Chemical Name or Material Potassium Iodide
Show More Show Less
Hazard Statement P305 + P351 + P338
Precautionary Statement H302-H315-H319
EINECSNumber 231-659-4
RTECSNumber TT2975000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.