Learn More
Description
Potassium Iodide USP comes as hexahedral crystals either transparent and colorless or somewhat opaque and white, or as a white, granular powder. It is slightly hygroscopic.
Specifications
Specifications
| Chemical Name or Material | Potassium Iodide USP |
| Packaging | Jar, 600mL, White, HDPE, Round, 89-400, TE Pano Neck |
| CAS | 7681-11-0 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 166 |
| Quantity | 500 g |
| Percent Purity | 99 to 101.5 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
