Learn More
Potassium Iodoatinate TS, (U.S.P. Test Solution), Spectrum™ Chemical
Shop All Spectrum Chemical Mfg. Corp. Products

Description
Spectrum™ Chemical Potassium Iodoplatinate TS, (U.S.P. Test Solution) is a solution, sometimes designated with 'TS or Test Solution', that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
Specifications
Specifications
| CAS Max % | 2, 97.6, 0.4 |
| Grade | Chemical Solution |
| Packaging | Amber Glass Bottle |
| Quantity | 50 mL |
| Chemical Name or Material | Potassium Iodoplatinate TS |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.