Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, Potassium iodomethyltrifluoroboronate, 95%, [888711-47-5], P10351 247.836 CH2BF3IK

Catalog No. 501455642
Encompass_Preferred

Compound: Potassium iodomethyltrifluoroboronate

CAS#: 888711-47-5

MDL#: not listed

Background: An organotrifluoroborate bearing a reactive iodomethyl group, used as a bifunctional building block in organic synthesis. The iodomethyl group is a versatile alkylating handle, while the trifluoroborate enables Suzuki-type coupling.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-145-5642 Supplier Frontier Specialty Chemicals Supplier No. P103511G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Potassium iodomethyltrifluoroboronate

CAS#: 888711-47-5

MDL#: not listed

Background: An organotrifluoroborate bearing a reactive iodomethyl group, used as a bifunctional building block in organic synthesis. The iodomethyl group is a versatile alkylating handle, while the trifluoroborate enables Suzuki-type coupling.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.