Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Potassium, Ion chromatography standard solution, Specpure™, K++ 1000μg/ml

Catalog No. AA47260AE
Change view
Click to view available options
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AA47260AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA47260AE Supplier Thermo Scientific™ Supplier No. 47260AE
Add to Cart

missing translation for 'validMainframeMsg'

KNO{3} in H{2}O

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula KNO{3} in H{2}O

Specifications

Physical Form Liquid
Quantity 100 mL
ShelfLife 1 Year 6 Months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.