Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Isopropylxanthate 98.0+%, TCI America™
SDP

Catalog No. I017725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I017725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I017725G Supplier TCI America Supplier No. I017725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 140-92-1
Molecular Formula C4H7KOS2
Molecular Weight (g/mol) 174.317
MDL Number MFCD00004930
InChI Key ZMWBGRXFDPJFGC-UHFFFAOYSA-M
Synonym Isopropylxanthic Acid Potassium Salt
PubChem CID 10931938
IUPAC Name potassium;propan-2-yloxymethanedithioate
SMILES CC(C)OC(=S)[S-].[K+]

Specifications

Quantity 25 g
Formula Weight 174.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Isopropylxanthate
EINECSNumber (2)-1787
RTECSNumber FG1581300
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.