Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassiu mL-Tartrate Monobasic

Catalog No. 243531100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-353-1100G 100 g
24-353-1500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-353-1100G Supplier Sigma Aldrich Supplier No. 243531100G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassiu mL-tartrate is found naturally in grapes. Its removal is necessary during wine making as it can precipitate, which is undesirable.Potassiu mL-tartrate monobasic may be used as a reductant during the synthesis of noble metal nanoparticles in the presence of linear polymers as protective agents.

Specifications

CAS 868-14-4
Density 1.954 g/mL at 25°C
Molecular Formula C4H5KO6
Linear Formula KO2CCH(OH)CH(OH)CO2H
MDL Number MFCD00065392
Quantity 100 g
Synonym KHT; L(+)-Tartaric acid monopotassium salt; Potassium bitartrate; Potassium hydrogen L-tartrate; Potassium hydrogen tartrate; Tartaric acid monopotassium salt
Molecular Weight (g/mol) 188.18 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.