Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassiu mL-Tartrate Monobasic

Catalog No. 255061KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 kg
6 x 1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-506-1KG 1 kg
25-506-6X1KG 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-506-1KG Supplier Sigma Aldrich Supplier No. 255061KG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassiu mL-tartrate monobasic can be used as a reducing agent to synthesize:Gold, palladium, silver and platinum nanoparticles from their salts. Polyvinylpyrrolidone capped platinum (PVP-Pt) nanoparticles from chloroplatinic acid and PVP.

Specifications

CAS 868-14-4
Molecular Formula C4H5KO6
Linear Formula C4H5KO6
MDL Number MFCD00065392
Quantity 1 kg
Synonym Potassium bitartrate; L-(+)-Tartaric acid monopotassium salt; Potassium hydrogen L-tartrate
Molecular Weight (g/mol) 188.18 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.