Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium Methoxide

Catalog No. 2927885G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-278-85G 5 g
29-278-8250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 29-278-85G Supplier Sigma Aldrich Supplier No. 2927885G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium methoxide can be used:As a methoxylating agent for methoxylation of bromo- and mesyloxyalkanes in the presence of an ionic liquid [bmim][BF4].As a catalyst for the synthesis of dimethyl carbonate and methyl formate.As a catalyst for the transesterification of sunflower oil for the production of biodiesel.

Specifications

CAS 865-33-8
Molecular Formula CH3KO
Linear Formula CH3OK
MDL Number MFCD00012416
Quantity 5 g
Synonym Potassium methylate
Molecular Weight (g/mol) 70.13 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.