Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium Methoxide Solution

Catalog No. 604021LF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-402-1LF 1 L
60-402-250MLF 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 60-402-1LF Supplier Sigma Aldrich Supplier No. 604021LF
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium methoxide solution in methanol has been used as a base to synthesize imines via aerobic oxidative coupling of benzylic alcohols and amines in the presence of gold nanoparticles supported on titanium dioxide catalyst.

Specifications

CAS 865-33-8
Density 0.95 g/mL at 20°C
Molecular Formula CH3KO
Refractive Index n20/D 1.37
Linear Formula CH3OK
MDL Number MFCD00012416
Quantity 1 L
Synonym Potassium methylate
Molecular Weight (g/mol) 70.13 g/mol
Biological Activity Eyes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.