Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Methyl Sulfate, MP Biomedicals™

Catalog No. MP521548110 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP521548110 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP521548110 Supplier MP Biomedicals, Inc Supplier No. 0521548110
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potassium Methyl Sulfate, White Crystalline Powder

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 562-54-9
Molecular Formula CH3KO4S
Molecular Weight (g/mol) 150.19
MDL Number MFCD00040472
InChI Key WBGWGHYJIFOATF-UHFFFAOYSA-M
Synonym Methylsulfuric acid potassium salt, Methyl sulfate potassium salt
IUPAC Name potassium methyl sulfate
SMILES [K+].COS([O-])(=O)=O

Specifications

Melting Point 217°C to 227°C
Color White
Quantity 10 g
Beilstein 3710745
Percent Purity 0.997
Physical Form Crystalline Powder
storageNote1 Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.