Learn More
Strem, An Ascensus Company CAS# 12030-85-2. 10g. Potassium niobate (99.999%-Nb) PURATREM. MFCD00049651
SDP
Supplier:  Strem, An Ascensus Company 93411710G
Encompass
CAS# 12030-85-2. 10g. Potassium niobate (99.999%-Nb) PURATREM. MFCD00049651. Molecular Weight: 180.01. Molecular Formula: KNbO3. Color/form: white pwdr. Strem# 93-4117. http://www.strem.com/catalog/v/93-4117/
Catalog No. 50-901-15303