Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Nitrate, 10% (w/v), pH 12, Ricca Chemical
SDP

Catalog No. 631916 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6319-16 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6319-16 Supplier Ricca Chemical Company Supplier No. 631916
Add to Cart
Add to Cart

Filling Solution for Cyanide ISE

Specifications

CAS 7757-79-1, 7732-18-5
Molecular Formula KNO3
MDL Number MFCD00011409
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
SMILES [K+].[O-][N+]([O-])=O
IUPAC Name potassium nitrate
Molecular Weight (g/mol) 101.10
PubChem CID 24434
ChEBI CHEBI:63043
Percent Purity 10%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 10% (w/v) Aqueous Solution
Packaging Natural Poly Bottle
pH 12
Quantity 500 mL
Chemical Name or Material Potassium Nitrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.