Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Nitrate ACS AR Crystal, Macron™

Catalog No. MK7028500 Shop All Avantor Products
Change view
Click to view available options
POLYSTORMOR; 12 x 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-7028-500 POLYSTORMOR; 12 x 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-7028-500 Supplier Avantor Supplier No. 702804

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

CAS 7757-79-1
Physical Form Crystalline
Quantity 12 x 500 g
Molecular Formula KNO3
MDL Number MFCD00011409
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
SMILES [K+].[O-][N+]([O-])=O
IUPAC Name potassium nitrate
Molecular Weight (g/mol) 101.10
PubChem CID 24434
ChEBI CHEBI:63043
Formula Weight 101.1
Grade ACS
Packaging POLYSTORMOR
Chemical Name or Material Potassium Nitrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.