Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Nitrite, ACS Reagent, Reagents

Catalog No. 01337819
Change view
Click to view available options
Quantity: 100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-337-819 Quantity: 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-819 Supplier Reagents Holdings Llc Supplier No. C1287000100C2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potassium Nitrite, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7758-09-0
Molecular Formula KNO2
Molecular Weight (g/mol) 85.10
InChI Key BXNHTSHTPBPRFX-UHFFFAOYSA-M
Synonym Nitrous Acid Potassium Salt
IUPAC Name potassium nitrite
SMILES [K+].[O-]N=O

Specifications

Boiling Point 537°C (may explode)
Color White or Slightly Yellow
Physical Form Solid
Quantity 100 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.915
Chemical Name or Material Potassium Nitrite, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.