Learn More
Potassium orthovanadate, 99.9% (metals basis)
Description
It is employed as in chemical fertilizer industries and used as corrosion inhibitor. It is also used for ammonia synthesis. Used as a catalyst in oil & gas industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Granules |
| Quantity | 25 g |
| UN Number | UN3285 |
| Solubility Information | Soluble in water. |
| Formula Weight | 232.24 |
| Percent Purity | 99.9% |
| Odor | Odorless |
| Assay | (metals basis) |
| Chemical Name or Material | Potassium orthovanadate |
Safety and Handling
| Hazard Category | H315-H319-H332-H335 |
| Hazard Statement | GHS H Statement H331-H315-H319-H335 Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S. |
| EINECSNumber | 238-220-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.