Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium orthovanadate, 99.9% (metals basis)

Catalog No. AA3950222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3950222 100 g
AA3950214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3950222 Supplier Thermo Scientific Chemicals Supplier No. 03950222
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 14293-78-8 | K3O4V | 232.232 g/mol

It is employed as in chemical fertilizer industries and used as corrosion inhibitor. It is also used for ammonia synthesis. Used as a catalyst in oil & gas industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14293-78-8
Molecular Formula K3O4V
Molecular Weight (g/mol) 232.232
MDL Number MFCD00210642
InChI Key FBRINGOENSVSPU-UHFFFAOYSA-N
Synonym Potassium vanadate (ortho)
PubChem CID 22023817
IUPAC Name tripotassium;trioxido(oxo)vanadium
SMILES [O-][V](=O)([O-])[O-].[K+].[K+].[K+]

Specifications

Physical Form Granules
Quantity 100 g
UN Number UN3285
Solubility Information Soluble in water.
Formula Weight 232.24
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Potassium orthovanadate
Hazard Category H315-H319-H332-H335
Hazard Statement GHS H Statement
H331-H315-H319-H335
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
EINECSNumber 238-220-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.